Learn More
Sigma Aldrich Fine Chemicals Biosciences Pefabloc SC, 100 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11429868001
Encompass
Synonym: 4-(2-aminoethyl)-benzene-sulfonyl fluoride, aebsf, aminoethyl-benzene-sulfonyl fluoride, 4-2-, proteinase k inhibitor. Pefabloc SC is used to inhibit the detrimental effects of proteases during preparative protein purification. Due to its low toxicity toward eukaryotic cells, it may be applied in the production of recombinant proteins, during fermentation of transformed cells, where proteolytic digestion may decrease the yield of the desired product.
Catalog No. NC1239417
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.