Learn More
Ted Pella Inc PELCO Conductive Gold Paste, 2g
Supplier: Ted Pella Inc 16022
Gold paste is fast drying and useful for analytical analysis over time where a high signal is desired. The gold is in microfine form and contains organic binders and a solvent. Dries at room temperature. Not intended for permanent use, but for testing and temporary work.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.