Learn More
3M™ PELTOR™ ComTac™ V Hearing Defender Headset
PELTOR™ ComTac™ V protects users from hazardous steady-state and impulse noise featuring a more natural sounding headset with excellent sound clarity.
Supplier: 3M™ 7100223428
Description
- Compatible with most helmets
- Dynamic Differential Microphone
- Level Dependent Function
- Noise canceling microphone
- Two-Way Communication
- Available in over-the-head or neckband styles
- Choose from three colors: Black, Olive Drab (O.D.) Green or Coyote Brown
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Olive drab green | |
Black | |
MT20H682BB-09 GN | |
22 dB | |
No | |
No |
Yes | |
Law Enforcement, Armed Forces | |
1 Ea. | |
Tactical Headsets | |
Neckband |