Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ PELTOR™ FLX2 Cable for CH-3 Headset With External PTT Button
PELTOR™ FLX2 platform allows workers to pair protective communication CH-3 two-way headsets with an external push-to-talk button.
Specifications
Color | Black |
---|---|
For Use With (Equipment) | For use with CH-3 Headsets with external PTT Button |
For Use With (Application) | Enables connection to two-way communication for CH-3 Headsets |
Pairs per Package | 1 Ea. |
Catalog Number | Mfr. No. | Model | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Product Type | Price | Quantity & Availability | ||||
19-072-290
|
3M™
7100185799 |
FLX2-200 | FLX2 Cable J11 | N/A | N/A | N/A | |||
19-072-244
|
3M™
7100207323 |
FLX2-201 | FLX2 Airline Cable | N/A | N/A | N/A | |||
Description
- For use with CH-3 Headset with external PTT Button
- J11 Plug Type
- Coiled polyurethane cable
- Aramid fiber reinforced
- Robust and highly flexible
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Black | |
For use with CH-3 Headsets with external PTT Button | |
Enables connection to two-way communication for CH-3 Headsets | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title