Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ ProTac III Headset Hearing Protector

Helps protect against hazardous and nuisance noise, while allowing environmental sounds to be reproduced by the electronic circuit, limited to 82dB

$1206.00

Specifications

Height (Metric) 200 mm
Width (Metric) 110 mm
View More Specs

Products 1
Catalog Number Mfr. No. dimIncludes Price Quantity & Availability  
Catalog Number Mfr. No. dimIncludes Price Quantity & Availability  
19-072-175
View Documents
3M™
7100088423
Case of 10 for $1,206.00
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

  • Professional-Quality Hearing Protection
  • Helps protect hearing from impulse noise and increases the ability to communicate with nearby colleagues, as well as hear warning signals, approaching vehicles, and sounds in machines and processes.
  • Environmental microphones for level-dependent hearing protection and ambient listening.
  • 3.5 mm listen-only stereo input for connection to external devices (e.g. cell phone, two-way radio, Ipod).
  • Voice guided menu system for ease of use and adjusting settings.
  • Auto power off, the headset will turn off after 4 hrs of non-use to save battery.
  • Low-battery warning to inform you when to change batteries
  • To reduce corrosion caused by sweat and moisture, the electronics are located in a separate compartment of the cup
  • Available in both in headband and hard hat attached versions (black).
  • Occupational use only.
Specifications

Specifications

200 mm
110 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.