Learn More
3M™ PELTOR™ WS™ ALERT™ XP Headset
PELTOR™ WS™ ALERT™ XP Headset protects users against hazardous noise, but also makes it easy to make phone calls with both hands free in noisy environments.
Specifications
Color | Black |
---|---|
ANSI S3.1974 | Yes |
Cushion Color | Black |
For Use With (Application) | Maintenance and Manufacturing |
Pairs per Package | 1 Ea. |
Catalog Number | Mfr. No. | Model | Style | Noise Reduction Rating (NRR) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Style | Noise Reduction Rating (NRR) | Price | Quantity & Availability | |||
19-040-626
|
3M™
7100147166 |
MRX21A2WS6-NA | Over-the-head | 23 dB |
Case for $571.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
19-040-630
|
3M™
7100147152 |
MRX21P3E2WS6-NA | Hard Hat | 24 dB |
Case for $512.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- High attenuating hearing protector available in two styles - stand-alone headband or safety helmet attachment
- Bluetooth™ technology for connection to one external device for hands-free talk and streaming
- Noise canceling boom microphone for clear speech transmission
- Level-dependent function for ambient listening to help improve situational awareness
- Auto-pairing to quickly connect to mobile devices
- Built-in FM-Radio with radio volume limited to 82dB
- Voice guided menu system
- Digital station search with ability to store up to 4 favorite stations
- Auto power will turn off after 4 hrs of non-use to save battery
- Low-battery warning at low battery level
Compliance
3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Black | |
Black | |
1 Ea. | |
No |
Yes | |
Maintenance and Manufacturing | |
Protective Communication |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.