Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ WS™ LiteCom PRO III Headset

A hearing protector with built-in analog and digital programmable two-way communication radio with level-dependent hearing protection.

$1604.40 - $9782.87

Specifications

Height (Metric) 224 mm
Width (Metric) 155 mm
View More Specs

Products 4
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
19-072-177
View Documents
3M™
7100099790
Headband with Bluetooth™ and a jack to connect to external devices
Case of 5 for $8,019.91
missing translation for 'QuantityLeft'
Add to Cart
 
19-072-178
View Documents
3M™
7100099586
Hard Hat Attached with Bluetooth™ and a jack to connect to external devices
Each for $1,604.40
Case of 5 EA for $8,022.01
missing translation for 'QuantityLeft'
Add to Cart
 
19-072-179
View Documents
3M™
7100099585
Headband with Bluetooth™
Case of 5 for $9,782.87
missing translation for 'QuantityLeft'
Add to Cart
 
19-072-180
View Documents
3M™
7100099785
Hard Hat Attached with Bluetooth™
Case of 5 for $9,634.82
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

  • Built-in programmable two-way communication radio allows for clear, hands-free communication in high levels of noise without cumbersome cables or cords.
  • The headset can be programmed to use up to 70 unique two-way radio channels using analog and/or DMR digital modulation.
  • Simultaneous, seamless connection to multiple external devices using Bluetooth™ technology.
  • Noise attenuating ear cups help provide hearing protection in environments with potentially hazardous noise.
  • Environmental listening with level dependent hearing protection for face-to-face communication and situational awareness.
  • Noise-cancelling waterproof speech microphone with voice operated transmission (VOX) for hands-free operation; helps provide clear communication in noisy and demanding environments.
  • User-friendly controls with voice-guided menu.
  • Built tough for the working professional.
  • Headset is available in headband, neckband and hard hat attached configurations.
Specifications

Specifications

224 mm
155 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.