Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ Earmuffs, Yellow

3M™ PELTOR™ Earmuffs are our innovative hearing protectors with a modern slim cup design.

Supplier:  3M™ 7000104076

 View more versions of this product

Catalog No. 17-000-637


Description

Description

  • Reduces noise levels
  • Colour-coding makes it easy to identify the level of protection being worn
  • Slim cups offer improved compatibility with other safety equipment
  • Wide soft ear cushions help workers stay comfortable
  • Designed to optimise weight distribution and balance for outstanding comfort
  • ABS plastic cups are rigid, high strength and impact resistant
  • Thermoplastic polyurethane (TPU) soft outer cup coating protects from abrasion and bumps

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Black, Yellow
Class A
Fits a Wide Variety of Industrial Hard Hats
X2P3E
24 dB
No
Yes
Black
Commercial Buildings, Design and Construction, Heavy Infrastructure, Industrial Maintenance
1 Ea.
X2 Series
Hard Hat
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.