Learn More
Enterprise Technology Solutions Maxtec MAX-13 Oxygen Cell is an FiO2 (Fraction of Inspired Oxygen) Sensor
SDP
Supplier:  Enterprise Technology Solutions CASG0030
Encompass_Preferred
The Maxtec MAX-13 Oxygen Cell is an FiO2 (Fraction of Inspired Oxygen) Sensor that is primarily used with ventilator machines. Check below for specific compatibility information.#G0-030 (Generic Equivalent):Response Time: Less than 12 seconds.Connector Type: 3.5 mm Mono Jack.Compatible With:BMD (Bio-Med Devices): M-10, M-2.Bird: 6400, 8400, VIP, Sentry.Covidien-Puritan Bennett: 7820, PB 850.Datex-Ohmeda: 7200, Engstrom Elvira, Engstrom Erica.MSA: MiniOx III.Respironics: Esprit Lifecare.
Catalog No. 50-240-7085
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.