Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentaerythrityl Tetraiodide 95.0+%, TCI America™
SDP

Catalog No. P073710G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 1522-88-9
Molecular Formula C5H8I4
Molecular Weight (g/mol) 575.737
MDL Number MFCD00019034
InChI Key HPMGSGCDKVCZHC-UHFFFAOYSA-N
Synonym 1,3-Diiodo-2,2-bis(iodomethyl)propane, Tetrakis(iodomethyl)methane
PubChem CID 73707
IUPAC Name 1,3-diiodo-2,2-bis(iodomethyl)propane
SMILES C(C(CI)(CI)CI)I

Specifications

Melting Point 227°C
Color White
Quantity 10 g
Formula Weight 575.74
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pentaerythrityl Tetraiodide
RTECSNumber TY1100000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.