Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Pentafluoroethyl)trimethylsilane, 97%, Thermo Scientific Chemicals

Catalog No. AC459300250
Click to view available options
Quantity:

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

124898-13-1

Molecular Formula

C5H9F5Si

Molecular Weight (g/mol)

192.2

InChI Key

MTPVUVINMAGMJL-UHFFFAOYSA-N

Synonym

pentafluoroethyl trimethylsilane, trimethyl pentafluoroethyl silane, pentafluoroethyltrimethylsilane, trimethylpentafluoroethylsilane, trimethyl perfluoroethyl silane, trimethyl 1,1,2,2,2-pentafluoroethyl silane, silane, trimethyl pentafluoroethyl, silane,trimethyl 1,1,2,2,2-pentafluoroethyl, acmc-20aple, trimethylsilylpentafluoroethane

PubChem CID

2760820

IUPAC Name

trimethyl(1,1,2,2,2-pentafluoroethyl)silane

SMILES

C[Si](C)(C)C(C(F)(F)F)(F)F

Specifications

Density

1.0500g/mL

Boiling Point

70.0°C

Infrared Spectrum

Authentic

Assay Percent Range

96% min. (GC)

Linear Formula

F3CCF2Si(CH3)3

Quantity

25 g

Specific Gravity

1.05

Solubility Information

Solubility in water: immiscible.

Formula Weight

192.2

Percent Purity

97%

Chemical Name or Material

(Pentafluoroethyl)trimethylsilane

Hazard Category

GHS Signal Word: Danger

Hazard Statement

GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Precautionary Statement

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.