Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Pentafluoroethyl)trimethylsilane, 97%, Thermo Scientific Chemicals

Catalog No. p-7077788
Click to view available options
Quantity:
1 g
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124898-13-1
Molecular Formula C5H9F5Si
Molecular Weight (g/mol) 192.2
InChI Key MTPVUVINMAGMJL-UHFFFAOYSA-N
Synonym pentafluoroethyl trimethylsilane, trimethyl pentafluoroethyl silane, pentafluoroethyltrimethylsilane, trimethylpentafluoroethylsilane, trimethyl perfluoroethyl silane, trimethyl 1,1,2,2,2-pentafluoroethyl silane, silane, trimethyl pentafluoroethyl, silane,trimethyl 1,1,2,2,2-pentafluoroethyl, acmc-20aple, trimethylsilylpentafluoroethane
PubChem CID 2760820
IUPAC Name trimethyl(1,1,2,2,2-pentafluoroethyl)silane
SMILES C[Si](C)(C)C(C(F)(F)F)(F)F

Specifications

Density 1.0500g/mL
Boiling Point 70.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula F3CCF2Si(CH3)3
Quantity 1 g
Specific Gravity 1.05
Solubility Information Solubility in water: immiscible.
Formula Weight 192.2
Percent Purity 97%
Chemical Name or Material (Pentafluoroethyl)trimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.