Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorophenyl 5-pyrid-3-ylthiophene-2-carboxylate, 97%, Thermo Scientific™

Catalog No. p-3586890
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 941716-94-5
Molecular Formula C16H6F5NO2S
Molecular Weight (g/mol) 371.28
MDL Number MFCD09966156
InChI Key IDMGFJPLJHXOJV-UHFFFAOYSA-N
Synonym pentafluorophenyl 5-pyrid-3-ylthiophene-2-carboxylate, pentafluorophenyl 5-pyridin-3-ylthiophene-2-carboxylate, pentafluorophenyl 5-pyridin-3-yl thiophene-2-carboxylate, 2,3,4,5,6-pentafluorophenyl 5-pyridin-3-yl thiophene-2-carboxylate, perfluorophenyl 5-pyridin-3-yl thiophene-2-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 5-pyridin-3-ylthiophene-2-carboxylate, 5-3-pyridinyl-2-thiophenecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 43811045
IUPAC Name 2,3,4,5,6-pentafluorophenyl 5-(pyridin-3-yl)thiophene-2-carboxylate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=C(S2)C2=CN=CC=C2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 371.28
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 5-pyrid-3-ylthiophene-2-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.