Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoropropionamide 98.0+%, TCI America™
SDP

Catalog No. p-7135072 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 354-76-7
Molecular Formula C3H2F5NO
Molecular Weight (g/mol) 163.05
MDL Number MFCD00039771
InChI Key KQTOYEUYHXUEDB-UHFFFAOYSA-N
Synonym pentafluoropropionamide, pentafluoropropanamide, propanamide, 2,2,3,3,3-pentafluoro, 2,2,3,3,3-pentafluoropropionamide, acmc-1cjql, pentafluoro propionic acid amide, 2,2,3,3,3-pentafluoropropionamide,, propanamide,2,2,3,3,3-pentafluoro
PubChem CID 67722
IUPAC Name pentafluoropropanamide
SMILES NC(=O)C(F)(F)C(F)(F)F

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 5 g
Formula Weight 163.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluoropropionamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.