Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAA1279136
Click to view available options
Quantity:

Pentafluoropropionic acid is used as an efficient catalyst for the preparation of dibenzo[a, j] xanthenes by condensation reaction of beta-naphthol with aryl aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluoropropionic acid is used as an efficient catalyst for the preparation of dibenzo[a, j] xanthenes by condensation reaction of beta-naphthol with aryl aldehydes.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents and strong bases.

Chemical Identifiers

CAS

422-64-0

Molecular Formula

C3HF5O2

Molecular Weight (g/mol)

164.03

MDL Number

MFCD00004170

InChI Key

LRMSQVBRUNSOJL-UHFFFAOYSA-N

Synonym

pentafluoropropionic acid, perfluoropropionic acid, pentafluoropropanoic acid, propanoic acid, pentafluoro, propionic acid, pentafluoro, 2,2,3,3,3-pentafluoropropionic acid, ipc-pffa-3, perfluoropropanoic acid, propanoic acid, 2,2,3,3,3-pentafluoro, perfluoropropionicacid

PubChem CID

62356

IUPAC Name

2,2,3,3,3-pentafluoropropanoic acid

SMILES

OC(=O)C(F)(F)C(F)(F)F

Specifications

Density

1.576

Boiling Point

95°C to 97°C

Odor

Rancid

Refractive Index

1.284

Quantity

500 g

UN Number

UN3265

Beilstein

1773387

Sensitivity

Hygroscopic

Formula Weight

164.03

Percent Purity

97%

Chemical Name or Material

Pentafluoropropionic acid

Hazard Category

H314-H332-H335

Hazard Statement

GHS H Statement
H314-H318-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

Precautionary Statement

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

DOTInformation

Hazard Class: 8; Packaging Group: III

EINECSNumber

207-021-6

RTECSNumber

UF6475000

TSCA

Yes

Recommended Storage

Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.