Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluoropropionic anhydride, 98%, Thermo Scientific Chemicals

Catalog No. AAA1447222
Click to view available options
Quantity:

Pentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.

Solubility
Reacts with water.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Chemical Identifiers

CAS

356-42-3

Molecular Formula

C6F10O3

Molecular Weight (g/mol)

310.047

MDL Number

MFCD00000429

InChI Key

XETRHNFRKCNWAJ-UHFFFAOYSA-N

Synonym

pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride

PubChem CID

67742

IUPAC Name

2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate

SMILES

C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F

Specifications

Density

1.589

Boiling Point

73°C to 74°C

Quantity

100 g

UN Number

UN3265

Beilstein

1806446

Sensitivity

Moisture sensitive

Solubility Information

Reacts with water.

Formula Weight

310.04

Percent Purity

98%

Chemical Name or Material

Pentafluoropropionic anhydride

Hazard Category

H314-H332-H335

Hazard Statement

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

Precautionary Statement

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

DOTInformation

Hazard Class: 8; Packaging Group: III

EINECSNumber

206-604-2

TSCA

Yes

Recommended Storage

Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.