Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane 97.0+%, TCI America™
SDP

Catalog No. p-7135609 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mL
500 mL

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Color Colorless
Boiling Point 36°C
Quantity 100 mL
Physical Form Clear Liquid at 20°C
UN Number 1265
Formula Weight 72.15
Percent Purity ≥97.0% (GC)
Chemical Name or Material Pentane
EINECSNumber (2)-0005
RTECSNumber RZ9450000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.