Learn More
TARGETMOL CHEMICALS INC PENTOSTATIN 25MG
Supplier:  TARGETMOL CHEMICALS INC T4006
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pentostatin (CI-825) is an extremely effective and irreversible inhibitor of adenosine deaminase (Ki 2.5 pM). purity: 99%
Catalog No. 50-258-0005
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.