Learn More
Medchemexpress LLC HY-A0006 50mg , Pentostatin Deoxycoformycin CAS:53910-25-1 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYA000650MG
Encompass_Preferred
Medchemexpress, HY-A0006 50mg Pentostatin Deoxycoformycin CAS:53910-25-1 Formula:C11H16N4O4 Ki: 2.5 pM (adenosine deaminase) Purity:98% Pentostatin is an irreversible inhibitor of adenosine deaminase with K i of 2.5 pM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3798
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.