Learn More
CPC Scientific H-Ala-Cys-Gly-Glu-Met-Gly-Trp-Val-Arg-Cys-Gly-Gly-Gly-Ser-C6-Lys-Lys(NOTA)-NH2 (trifluoroacetate salt) 1MG
Supplier: CPC Scientific CANS012A
Metal chelating peptide Pep-1L-NOTA targets tumor-restricted receptor, IL13RA2, found to be present in some aggressive malignancies, specifically gliomas and malignant melanoma. Recent work has demonstrated that the copper-bound peptide selectively binds hIL13RA2-expressing tumors and validates its use in peptide-based PET imaging for cancer diagnostics.ONE-LETTER SEQUENCE: ACGEMGWVRCGGGS-C6-K-K(NOTA)-NH2MOLECULAR FORMULA: C84H139N27O25S3MOLECULAR WEIGHT:2023.36STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Sai, Kiran Kumar Solingapuram, et al. "Peptide-based PET imaging of the tumor restricted IL13RA2 biomarker." Oncotarget 5 (2017).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.