Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peppermint Oil, NF, 5%, Spectrum™ Chemical
SDP

Catalog No. 18605009 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 mL
Packaging:
Amber Glass Bottle

PE105, 8006-90-4

Spectrum™ Chemical Peppermint Oil, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS 8006-90-4
CAS Max % 1
Packaging Amber Glass Bottle
Refractive Index 1.459-1.465
Quantity 25 mL
Percent Purity 5%
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.