Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peppermint Oil, Rectified, FCC, 5%, Spectrum™ Chemical
SDP

Catalog No. p-7157959 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
10 kg
Packaging:
Amber Glass Bottle
Steel Pail

P1595, 8006-90-4

Spectrum™ Chemical Peppermint Oil, Rectified, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8006-90-4

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Refractive Index 1.459-1.465
Quantity 25 mL
Specific Gravity 0.896-0.908
Percent Purity 5%
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.