Learn More
Gold Biotechnology Inc Pepstatin A 25 mg
Supplier: Gold Biotechnology Inc P02025
Pepstatin A is a competitive, reversible aspartic protease inhibitor and inhibits proteases such as pepsin, chymosin, renin, HIV proteases, and cathepsins D and E. Peptstatin A is highly selective and is often used in a mixture with other enzyme inhibitors. In vitro studies have shown pepstatin A is the only aspartic acid inhibitor that exhibits inhibitory effects on HIV proteases.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.