Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perchloric Acid, 0.100 N (N/10) in Glacial Acetic Acid, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Safety-coated Glass Bottle
Specifications
Specifications
Color | Colorless to Yellow |
Concentration | 0.100N |
CAS Min % | 94.0, 2.33, 1.28 |
CAS Max % | 98.0, 2.43, 1.33 |
Packaging | Amber Glass Bottle |
Solvent or Matrix | Glacial Acetic Acid |
Quantity | 500 mL |
Physical Form | Liquid |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction