Learn More
GFS Chemicals Inc PERCHLORIC ACID, 70%, VERITAS DOUBLE DISTILLED | 500 ML | GFS CHEMICALS, INC. Item #: 55802 | Bulk #: 230
SDP
Supplier:  GFS Chemicals Inc 55802
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
PERCHLORIC ACID, 70%, VERITAS DOUBLE DISTILLED | Chemical Formula : HClO!4 | CAS # : 7601-90-3 | Specifications : Ag (ppt), Al (ppt), As (ppt), Assay 69.0 - 72.0 %, Au (ppt), Ba (ppt), Be (ppt), Bi (ppt), Ca (ppt), Cd (ppt), Chloride 0.00001 % , Co (ppt), Cr (ppt), Cu (ppt), Fe (ppt), Ga (ppt), Ge (ppt), Heavy Metals 0.000001 % , Hg (ppt), In (ppt), Iron 0.000001 % , K (ppt), La (ppt), Li (ppt), Mg (ppt), Mn (ppt), Mo (ppt), Na (ppt), Nb (ppt), Ni (ppt), Nitrogen compounds (%) 0.0001 % , Pb (ppt), Pd (ppt), Pt (ppt), Residue after ignition 0.0001 % , Sb (ppt), Silicate and phosphate (%) 0.00001 % , Sn (ppt), Sr (ppt), Sulfate 0.0001 % , Ta (ppt), Ti (ppt), Tl (ppt), V (ppt), W (ppt), Zn (ppt), Zr (ppt)
Catalog No. NC9780926
missing translation for 'QuantityLeft'