Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, in Glacial Acetic Acid, for Non-Aqueous Titrations, Certified, 0.1000N ±0.0005N, LabChem™

Catalog No. p-5388863
Click to view available options
Quantity:
500mL
1L
2.5L
4L
Packaging:
Amber Glass
Clear Glass

Chemical Identifiers

CAS 64-19-7, 108-24-7, 7601-90-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.052
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(=O)O

Specifications

CAS Max % 97.04, 2.1, 0.86
Linear Formula HClO4
UN Number UN2789
Concentration 0.1000N ±0.0005N
Density 1.06g/mL
Formula Weight 100.47
Physical Form Liquid
Grade Certified
Identification Passes Test
Odor Vinegar
Packaging Amber Glass
Solubility Soluble in water
Specific Gravity 1.06g/mL
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Flash Point 40°C
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases; Thermal decomposition generates: Corrosive vapors
Color Colorless to Yellow
Quantity 500mL
Chemical Name or Material Perchloric Acid, in Glacial Acetic Acid, for Non-Aqueous Titrations
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minut
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.