Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Periodic Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Specifications
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 100 g |
IUPAC Name | pentahydroxy(oxo)-$l^{7}-iodane |
Formula Weight | 227.96 |
Grade | BAKER ANALYZED™ ACS Reagent |
Chemical Name or Material | Periodic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction