Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periodic Acid, MP Biomedicals™

Catalog No. p-7083106 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
25 g
100 g
500 g

Used for cell staining. Periodic Acid, MP Biomedicals stains the cytoplasm of cells found in the tubular pattern, mucus, and apical surfaces of various lumen and glycoproteins on SDS-PAGE gels. This compound can be used in oxidation reactions.

  • Periodic acid can be used in oxidation reactions converting sulfides into sulfones, catalyzed by Chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones
  • Periodic acid can also react with the 1,2 glycol linkage of carbohydrates forming aldehydes in tissue sections, which can be colored with Schiff bases, thus allowing for the staining of these tissues
  • Furthermore, studies show that periodic acid is not effective in staining cancer cells in solid nests, which are better stained using Grimelius

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
IUPAC Name tetrahydroxyiodous acid
SMILES O[I](O)(O)(O)(O)=O

Specifications

Melting Point 122°C (Lit.)
Boiling Point 135°C (literature)
Quantity 25 g
Physical Form Yellowish Crystalline Powder
Solubility Information Freely soluble in water (100mg/mL), alcohol, nitric acid; slightly soluble in ether.
Formula Weight 227.94
Percent Purity ≥99%
Recommended Storage Room Temperature (15–30°C). Store dessicated.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.