Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Prohexadione-Calcium, MilliporeSigma™ Supelco™

Catalog No. 111012892 Shop All MilliporeSigma Supelco Products
Click to view available options
:

Prohexadione-Calcium (Pro-Ca, prohexadione-Ca or BX-112) is a plant growth retardant which is used to inhibit gibberellin biosynthesis of plants. It generally reduces shoot elongation in plants.

Agriculture

Specifications

Quantity

100 mg

CAS

127277-53-6

Type

Analytical Standard

Product Line

PESTANAL

Linear Formula

C10H10 CaO5

MDL Number

MFCD03095704

Synonym

3,5-Dioxo-4-propionyl-cyclohexanecarboxylic acid calcium salt

Formula Weight

250.26

Grade

Analytical Standard

Physical Form

Neat

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.