Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PESTANAL™ TCPP, Mixture of isomers, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
:
100 mg
Description
Agriculture
Specifications
Specifications
Quantity | 100 mg |
CAS | 13674-84-5 |
Type | Analytical Standard |
Product Line | PESTANAL |
Linear Formula | C9H18 Cl3O4 P |
Formula Weight | 327.57 |
Grade | Analytical Standard |
Flash Point | 185°C - Closed Cup |
Physical Form | Neat |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction