Learn More
TARGETMOL CHEMICALS INC PF-00217830 5MG
Supplier:  TARGETMOL CHEMICALS INC T24613
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PF-00217830 is a serotonin 1A receptor agonist dopamine D2 receptor agonist and serotonin 2A receptor antagonist.PF-00217830 may be used in the study of schizophrenia. purity: 98%
Catalog No. 50-258-0793
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.