Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PF 4800567 hydrochloride, Tocris Bioscience™

Catalog No. p-4816868 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective casein kinase 1ε inhibitor

Selective casein kinase 1ε inhibitor; displays 22-fold greater potency towards CK1ε than CK1δ (IC50 values are 32 and 711 nM for CK1ε and CK1δ respectively). ATP competitive. Displays minimal effect on the circadian clock.

Chemical Identifiers

CAS 1391052-28-0
Molecular Formula C17H19Cl2N5O2
Molecular Weight (g/mol) 396.272
InChI Key QZXZQMUZEHTFHD-UHFFFAOYSA-N
Synonym pf 4800567 hydrochloride, pf-4800567 hydrochloride, 3-3-chlorophenoxy methyl-1-tetrahydro-2h-pyran-4-yl-1h-pyrazolo 3,4-d pyrimidin-4-amine hydrochloride, 3-3-chlorophenoxymethyl-1-oxan-4-yl pyrazolo 3,4-d pyrimidin-4-amine hydrochloride
PubChem CID 71751553
IUPAC Name 3-[(3-chlorophenoxy)methyl]-1-(oxan-4-yl)pyrazolo[3,4-d]pyrimidin-4-amine;hydrochloride
SMILES C1COCCC1N2C3=C(C(=N2)COC4=CC(=CC=C4)Cl)C(=NC=N3)N.Cl

Specifications

Quantity 10 mg
Formula Weight 396.27
Percent Purity >99%
Chemical Name or Material PF 4800567 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.