Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFI 1, Tocris Bioscience™

Catalog No. p-4817136 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

BET bromodomain inhibitor

Potent BET bromodomain inhibitor; exhibits inhibitory activity at bromodomain-containing protein (BRD) 2 and BRD4 (IC50 values are 98 and 220 nM respectively). Induces apoptosis and G1 cell cycle arrest in BET inhibitor-sensitive cell lines (MV4;11). Also downregulates Aurora B expression in MV4;11 cells. Cell permeable.

Chemical Identifiers

CAS 1403764-72-6
Molecular Formula C16H17N3O4S
Molecular Weight (g/mol) 347.389
InChI Key TXZPMHLMPKIUGK-UHFFFAOYSA-N
Synonym pfi-1, 2-methoxy-n-3-methyl-2-oxo-1,2,3,4-tetrahydroquinazolin-6-yl benzenesulfonamide, 2-methoxy-n-3-methyl-2-oxo-1,2,3,4-tetrahydroquinazolin-6-yl benzene-1-sulfonamide, 2-methoxy-n-3-methyl-2-oxo-1,4-dihydroquinazolin-6-yl benzenesulfonamide, d03lnf, pfi 1, pfi hplc, 2-methoxy-n-3-methyl-2-oxo-1,2,3,4-tetrahydroquinazolin-6-yl benzenesulfon amide
PubChem CID 71271629
IUPAC Name 2-methoxy-N-(3-methyl-2-oxo-1,4-dihydroquinazolin-6-yl)benzenesulfonamide
SMILES CN1CC2=C(C=CC(=C2)NS(=O)(=O)C3=CC=CC=C3OC)NC1=O

Specifications

Quantity 10 mg
Formula Weight 347.39
Percent Purity >98%
Chemical Name or Material PFI 1
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.