Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PG 01037 dihydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Dopamine D3 receptor antagonist; 133-fold selective for D3 over D2 receptors in vitro (Ki values are 0.70, 93.3 and 375 nM for D3, D2 and D4 receptors respectively). Attenuates abnormal involuntary movements associated with L-DOPA (Cat. No. 3788) in rat models of Parkinson's disease. Inhibits the effects of methamphetamine; attenuates drug-induced behaviors in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 554.34 |
Percent Purity | >99% |
Chemical Name or Material | PG 01037 dihydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction