Learn More
Pharmacal Research Laboratories pH Control, 30 Gal
pH Control is specifically formulated for use where neutralization of acid materials is required prior to disposal. This product is also used as an intermediate compound between an acid and alkaline so that cleaning can be done in the same wash solution. This compound is stable in wash temperatures in excess of 180° F and will not break down under these conditions.