Learn More
CTL Scientific Supply Corp PH-FIX ST. 7.9-9.8,CS/10BX/100
Supplier: CTL Scientific Supply Corp 9217010
PH-FIX 7.9-9.8 TEST PAPER are non-bleeding indicator strips. Gradation of scale: 7.9 - 8.3 - 8.6 - 8.9 - 9.1 - 9.4 - 9.8 (case of 10 boxes of 100 strips each)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.