Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ pH Paper Rolls

General purpose pH papers in efficient 15' rolls
Description
- Convenient dispenser allows you to select the right amount of pH paper for each application
- Easy-to-usedash (em)Dip paper in solution to be tested and match to color chart immediately
- Highly accurate and well suited for most applications
- Each SKU is packed in box of five clear plastic dispensers
- Cat. No. 13-640-507 is a pH paper testing set that includes shrink wrapped pack of seven dispensers (one of each SKU)
- Store at room temperature. Keep out of direct sunlight.
Order Info
15 ft. Roll provides about 100 tests.
Specifications
3 to 5.5 | |
0.5 | |
Room Temperature |
Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications | |
100 Tests | |
pH Test Paper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction