Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHA 767491 hydrochloride, Tocris Bioscience™

Catalog No. p-4784283 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MK2 inhibitor; also cdk inhibitor; potently inhibits cdk9

ATP-competitive cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 0.034 - 0.75, 0.24 - 1.2, 0.25 - 1.1 and 0.46 - 1 μM for cdk9, cdk2, cdk1 and cdk5, respectively). Also inhibits mitogen-activated protein kinase-activated protein kinase-2 (MK2; IC50 = 171 nM). Prevents initiation of DNA replication. Inhibits cell proliferation in a variety of human cell lines and induces apoptosis in a p53-independent manner in vivo.

Chemical Identifiers

CAS 942425-68-5
Molecular Formula C12H12ClN3O
Molecular Weight (g/mol) 249.698
InChI Key IMVNFURYBZMFDZ-UHFFFAOYSA-N
Synonym pha 767491 hydrochloride, pha-767491 hydrochloride, 1,5,6,7-tetrahydro-2-4-pyridinyl-4h-pyrrolo 3,2-c pyridin-4-one hydrochloride, 2-pyridin-4-yl-1h,5h,6h,7h-pyrrolo 3,2-c pyridin-4-one hydrochloride, cay-10572 hydrochloride, pha-00767491 hydrochloride, pha 767491 hydrochloride hplc, 1,5,6,7-tetrahydro-2-4-pyridinyl-??4h-pyrrolo 3,2-c pyridin-4-one hydrochloride, 2-pyridin-4-yl-1,5,6,7-tetrahydro-4h-pyrrolo 3,2-c pyridin-4-one hydrochloride
PubChem CID 11715766
IUPAC Name 2-pyridin-4-yl-1,5,6,7-tetrahydropyrrolo[3,2-c]pyridin-4-one;hydrochloride
SMILES C1CNC(=O)C2=C1NC(=C2)C3=CC=NC=C3.Cl

Specifications

Quantity 10 mg
Formula Weight 249.7
Percent Purity >99%
Chemical Name or Material PHA 767491 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.