Learn More
Saint Gobain Performance Plastics PHARMED BPT PUMP TUBING 25FT
Supplier: Saint Gobain Performance Plastics AY242022
Saint-Gobain PharMed BPT Tubing Diameter Inner 5/16 in. 7.9 mm Diameter Outer 7/16 in. 11 mm Thickness 0.063 in. 1.6 mm Autoclavable Autoclavable Certifications/Compliance Fully tested to ISO 10993 standards. Meets USP Class VI FDA and NSF criteria.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.