Learn More
Saint Gobain Performance Plastics PHARMED BPT PUMP TUBING
Supplier:  Saint Gobain Performance Plastics AY242606
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Saint-Gobain PharMed BPT Tubing Diameter Inner 0.031 in. 0.8 mm Diameter Outer 0.16 in. 4 mm Thickness 0.063 in. 1.6 mm Autoclavable Autoclavable Certifications/Compliance Fully tested to ISO 10993 standards. Meets USP Class VI FDA and NSF criteria.
Catalog No. 50-206-9046