Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenethyl Isocyanate 98.0+%, TCI America™
SDP

Catalog No. p-7135202 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1943-82-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00037044
InChI Key HACRKYQRZABURO-UHFFFAOYSA-N
Synonym phenethyl isocyanate, 2-isocyanatoethyl benzene, phenylethyl isocyanate, 2-phenylethyl isocyanate, 2-phenylethylisocyanate, phenylethylisocyanate, phenethylisocyanate, benzene, 2-isocyanatoethyl, 2-isocyanato-ethyl-benzene, 2-phenethyl isocyanate
PubChem CID 160602
ChEBI CHEBI:60067
IUPAC Name 2-isocyanatoethylbenzene
SMILES C1=CC=C(C=C1)CCN=C=O

Specifications

Color Yellow
Boiling Point 210°C
Quantity 5 g
UN Number 2206
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenethyl Isocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.