Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl 3,4,6-Tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-beta-D-glucopyranoside 98.0+%, TCI America™
SDP

Catalog No. P18665G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

187022-49-7

Molecular Formula

C21H24Cl3NO9S

Molecular Weight (g/mol)

572.831

MDL Number

MFCD11112181

InChI Key

AZDNCEYOFMNMSH-QTMHVTGLSA-N

PubChem CID

44630021

IUPAC Name

[(2S,3S,4R,5S,6S)-3,4-diacetyloxy-6-phenylsulfanyl-5-(2,2,2-trichloroethoxycarbonylamino)oxan-2-yl]methyl acetate

SMILES

CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)NC(=O)OCC(Cl)(Cl)Cl)OC(=O)C)OC(=O)C

Specifications

Melting Point

143°C

Color

White

Quantity

5 g

Formula Weight

572.83

Percent Purity

≥98.0% (HPLC)

Physical Form

Crystalline Powder

Chemical Name or Material

Phenyl 3,4,6-Tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-beta-D-glucopyranoside

TSCA

No

Recommended Storage

Freezer

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.