Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetaldehyde, 99.5%, For GC analysis, MP Biomedicals™

Catalog No. p-4405186 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mL
500mL

Phenylacetaldehyde ≥95%, Clear Liquid

  • It is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions.
  • It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages.
  • It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin.
  • Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.
  • A naturally occurring aromatic compound. Contains varying amounts of poly(styrene oxide).

Chemical Identifiers

CAS 122-78-1
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00006993
InChI Key DTUQWGWMVIHBKE-UHFFFAOYSA-N
PubChem CID 998
ChEBI CHEBI:16424
IUPAC Name 2-phenylacetaldehyde
SMILES O=CCC1=CC=CC=C1

Specifications

Color Colorless
Density 1.027g/mL
Assay Percent Range 0.995
Quantity 100 mL
Synonym phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde
Formula Weight 120.15
Physical Form Liquid
Chemical Name or Material Phenylacetaldehyde, For GC analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.