Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylephrine Related Compound E Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018048 Shop All MilliporeSigma Supelco Products
Click to view available options
:
50 mg

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 71786-67-9
Quantity 50 mg
Type Certified Reference Material
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C16H17 NO2 · HCl
Synonym 1-(3-Hydroxyphenyl)-2-[methyl(phenylmethyl)amino]ethanone hydrochloride; Benzylphenylephrone; 2-[Benzyl(methyl)amino]-1-(3-hydroxyphenyl)ethanone hydrochloride; Benzylphenylephrone hydrochloride
Formula Weight 291.77
Grade Certified Reference Material
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.