Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylhydroquinone 98.0+%, TCI America™
SDP

Catalog No. p-7126301 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1079-21-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002342
InChI Key XCZKKZXWDBOGPA-UHFFFAOYSA-N
Synonym phenylhydroquinone, 2-phenylhydroquinone, 2,5-dihydroxybiphenyl, 1,1'-biphenyl-2,5-diol, biphenyl-2,5-diol, hydroquinone, phenyl, 1,4-benzenediol, phenyl, 2,5-biphenyldiol, o-phenylhydroquinone
PubChem CID 14116
IUPAC Name [1,1'-biphenyl]-2,5-diol
SMILES OC1=CC=C(O)C(=C1)C1=CC=CC=C1

Specifications

Melting Point 101°C
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylhydroquinone
EINECSNumber (4)-1336
RTECSNumber DV4550000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.