Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylsulfonylacetonitrile 98.0+%, TCI America™
SDP

Catalog No. p-7134773 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 7605-28-9
Molecular Formula C8H7NO2S
Molecular Weight (g/mol) 181.209
MDL Number MFCD00007550
InChI Key ZFCFFNGBCVAUDE-UHFFFAOYSA-N
Synonym benzenesulfonylacetonitrile, phenylsulfonyl acetonitrile, 2-phenylsulfonyl acetonitrile, phenylsulfonylacetonitrile, 2-benzenesulfonyl acetonitrile, phenylsulphonylacetonitrile, acetonitrile, phenylsulfonyl, benzenesulphonyl acetonitrile, benzenesulfonyl-acetonitrile, 2-phenylsulfonyl ethanenitrile
PubChem CID 82077
IUPAC Name 2-(benzenesulfonyl)acetonitrile
SMILES C1=CC=C(C=C1)S(=O)(=O)CC#N

Specifications

Melting Point 112°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 181.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylsulfonylacetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.