Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiohydantoin-DELTA-threonine 90.0+%, TCI America™
SDP

Catalog No. p-7134577 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 5800-50-0
Molecular Formula C11H10N2OS
Molecular Weight (g/mol) 218.274
MDL Number MFCD00070617
InChI Key SXYSWGDIVRMEDE-MBXJOHMKSA-N
Synonym 5-Ethylidene-3-phenyl-2-thiohydantoin, PTH-DELTA-threonine
PubChem CID 45105912
IUPAC Name (5Z)-5-ethylidene-3-phenyl-2-sulfanylideneimidazolidin-4-one
SMILES CC=C1C(=O)N(C(=S)N1)C2=CC=CC=C2

Specifications

Melting Point 236°C
Quantity 100 mg
Formula Weight 218.27
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-DELTA-threonine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.