Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiohydantoin-glycine 98.0+%, TCI America™
SDP

Catalog No. p-7134579 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 2010-15-3
Molecular Formula C9H8N2OS
Molecular Weight (g/mol) 192.24
MDL Number MFCD00027413
InChI Key ZZRIQDWDJVLELF-UHFFFAOYSA-N
Synonym pth-glycine, 3-phenyl-2-thiohydantoin, phenylthiohydantoin, 3-phenyl-2-thioxoimidazolidin-4-one, phenylthiohydantoin glycine, 3-phenyl-2-thioguidanthion, hydantoin, 3-phenyl-2-thio, glycine, 3-phenyl-2-thiohydantoin, 4-imidazolidinone, 3-phenyl-2-thioxo, 2-mercapto-3-phenyl-3,5-dihydro-4h-imidazol-4-one
PubChem CID 700731
IUPAC Name 3-phenyl-2-sulfanylideneimidazolidin-4-one
SMILES O=C1CNC(=S)N1C1=CC=CC=C1

Specifications

Quantity 100 mg
UN Number 2811
Formula Weight 192.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-glycine
RTECSNumber MU4025000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.