Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiohydantoin-(Nepsilon-phenylthiocarbamyl)-lysine 98.0+%, TCI America™
SDP

Catalog No. p-7134702 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 29635-94-7
Molecular Formula C20H22N4OS2
Molecular Weight (g/mol) 398.543
MDL Number MFCD00151476
InChI Key BUPYRERCBPFRFM-KRWDZBQOSA-N
Synonym PTH-(Nepsilon-PTC)-lysine
PubChem CID 1550115
IUPAC Name 1-[4-[(4S)-5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl]butyl]-3-phenylthiourea
SMILES C1=CC=C(C=C1)NC(=S)NCCCCC2C(=O)N(C(=S)N2)C3=CC=CC=C3

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 100 mg
Formula Weight 398.54
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-(Nepsilon-phenylthiocarbamyl)-lysine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.