Learn More
Phenyltin trichloride, Thermo Scientific Chemicals
Description
Phenyltin trichloride is used to produce 3-phenyl-cyclohexanone with cyclohex-2-enone. And it is also used in pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Density | 1.84 g/mL |
Odor | Pungent |
Quantity | 5 g |
UN Number | UN2788 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 302.18 |
Physical Form | Liquid |
Chemical Name or Material | Phenyltin trichloride |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H301-H311-H331-H314-H318 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 214-393-3 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.